16-O-Methylcafestol
Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing