Skip to Content
Merck
1-[2-(Trifluoromethyl)phenyl]imidazole

1-[2-(Trifluoromethyl)phenyl]imidazole

1-[2-(Trifluoromethyl)phenyl]imidazole
Synonym(s):
TRIM
Empirical Formula (Hill Notation):
C10H7F3N2
CAS Number:
Molecular Weight:
212.17

Products

Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing