Product Name
Oleandomycin phosphate salt ~95%
Product Number
o6125
Product Brand
sigma
CAS Number
7060-74-4
Molecular Weight
785.85
Test | Specification |
---|---|
APPEARANCE | WHITE TO YELLOW POWDER |
SOLUBILITY | CLEAR TO VERY SLIGHTLY HAZY YELLOW TO YELLOW-GREEN SOLUTION AT 100 MG/ML IN WATER |
CARBON | 50.8 TO 54.8% |
NITROGEN | 1.5 TO 2.1% |
ICP-ATOMIC EMISSION | 3.6 TO 4.2% PHOSPHORUS |
PURITY BY THIN LAYER CHROMATOGRAPHY | APPROX. 95% (92 TO 100%) |
SHELF LIFE | 6 YEARS |
DOCUMENT # O6125/09/21/98/1 |