Product Name
2′,4′,6′-Trihydroxyacetophenone monohydrate suitable for matrix substance for MALDI-MS, ≥99.5%
Product Number
91928
Product Brand
sial
CAS Number
249278-28-2
Molecular Weight
186.16
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals
TITRATION (NT) TBAOH 0.1M≥ 99.5 %
PURITY (HPLC AREA %)≥ 99.5 %
MELTING POINT218 - 222 C
WATER8 - 11 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
MISCELLANEOUS TESTSUV: CUT-OFF AT ≤ 400 NM (1 % IN ETHANOL)
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 200 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 50 mg/kg
MAGNESIUM (ICP)≤ 5 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
SODIUM (ICP)≤ 50 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
RECOMMENDED RETEST PERIOD54 MONTHS