Product Name
Dihydroxyfumaric acid hydrate 98%
Product Number
d113204
Product Brand
aldrich
CAS Number
199926-38-0
Molecular Weight
148.07 (anhydrous basis)
Test | Specification |
---|---|
APPEARANCE (COLOR) | Off White to Yellow and Faint Beige to Beige |
APPEARANCE (FORM) | Powder |
TITRATION (T) NAOH 0.1M | 97.5 - 102.5 % |
REMARKS ON TITRATION | ON DRIED BASIS |
LOSS ON DRYING | 5 - 20 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |