Product Name
2-Chloro-1,3,2-benzodioxaphosphorin-4-one 95%
Product Number
324124
Product Brand
aldrich
CAS Number
5381-99-7
Molecular Weight
202.53
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE OR LIGHT YELLOW CRYSTALS |
OR SOLID AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
AND/OR | |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 94.0-106.0% (WITH AGNO3 AFTER O2 COMBUSTION) |
REVISED JULY 19, 2006 RJM |