Skip to Content
Merck

Skip To

1613010

USP

Desiccant, Small

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Synthetic zeolite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 x 44 g
Please contact Customer Service for Availability
CZK 9,050.00

About This Item

Linear Formula:
Na4[(AlO2)4(SiO2)4] · 9H2O
CAS Number:
Molecular Weight:
730.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 9,050.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

packaging

pkg of 88 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

neat

SMILES string

[Si](=O)=O.[Al](=O)O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Desiccant, Small USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1613032NISTRM8850NIST8851
application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical (small molecules)

application(s)

-

application(s)

-

format

neat

format

neat

format

matrix material

format

-

packaging

pkg of 88 mg

packaging

pkg of 2 × 80 g

packaging

-

packaging

pkg of 35-40 g

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters