Skip to Content
Merck
All Photos(1)

Key Documents

Y0002175

Olanzapine embonate monohydrate

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Naphthalenecarboxylic acid, 4,4′-methylenebis[3-hydroxy-, compd. with 2-methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, hydrate (1:1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16O6 · C17H20N4S · H2O
CAS Number:
Molecular Weight:
718.82
UNSPSC Code:
41116107
NACRES:
NA.24

API family

olanzapine

manufacturer/tradename

EDQM

storage temp.

2-8°C

InChI

1S/C23H16O6.C17H20N4S.H2O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12;/h1-10,24-25H,11H2,(H,26,27)(H,28,29);3-6,11,19H,7-10H2,1-2H3;1H2

InChI key

ZIMCQJVMPKQQPB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Olanzapine embonate monohydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service