Skip to Content
Merck
All Photos(1)

Documents

T2299

Sigma-Aldrich

Thimerosal

~98% (HPLC)

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
EC Number:
MDL number:
UNSPSC Code:
51101500

Assay

~98% (HPLC)

mp

234-237 °C (dec.) (lit.)

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willeke M C van Roon-Mom et al.
BMC molecular biology, 9, 84-84 (2008-10-11)
Huntington's disease is a progressive autosomal dominant neurodegenerative disorder that is caused by a CAG repeat expansion in the HD or Huntington's disease gene. Although micro array studies on patient and animal tissue provide valuable information, the primary effect of
Yan Xu et al.
Antimicrobial agents and chemotherapy, 54(1), 536-539 (2009-10-21)
The in vitro activity of thimerosal versus those of amphotericin B and natamycin was assessed against 244 ocular fungal isolates. The activity of thimerosal against Fusarium spp., Aspergillus spp., and Alternaria alternata was 256 times, 512 times, and 128 times

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service