Skip to Content
Merck
All Photos(1)

Key Documents

SML1039

Sigma-Aldrich

ENMD-1068 hydrochloride

≥97% (HPLC)

Synonym(s):

6-Amino-1-[4-(3-methyl-1-oxobutyl)-1-piperazinyl]-1-hexanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29N3O2 · HCl
CAS Number:
Molecular Weight:
319.87
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C15H29N3O2/c1-13(2)12-15(20)18-10-8-17(9-11-18)14(19)6-4-3-5-7-16/h13H,3-12,16H2,1-2H3

InChI key

VRZYMMVEALDDBH-UHFFFAOYSA-N

Application

ENMD-1068 hydrochloride has been used as a protease-activated receptor 2 (PAR2) antagonist to determine the pathway responsible for paw edema induced by Crotalus durissus terrificus venom serine protease (Cdtsp 2).

Biochem/physiol Actions

ENMD-1068 hydrochloride exhibits anti-angiogenic and anti-inflammatory activities.
ENMD-1068 is an antagonist of protease-activated receptor 2 (PAR2). The compound blocks TNFα production in synovial explants from patients with arthritis, and inhibits mast cell tryptase-induced recruitment of eosinophils into the pleural cavity of mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline R C Costa et al.
International journal of molecular sciences, 19(8) (2018-08-17)
Snake venom serine proteases (SVSPs) represent an essential group of enzymatic toxins involved in several pathophysiological effects on blood homeostasis. Some findings suggest the involvement of this class of enzymatic toxins in inflammation. In this paper, we purified and isolated
Yifeng Wang et al.
American journal of obstetrics and gynecology, 210(6), 531-531 (2014-02-06)
Protease-activated receptor 2 plays an important role in the pathogenesis of endometriosis. We studied the effect of ENMD-1068, a protease-activated receptor 2 antagonist, on the development of endometriosis in a noninvasive fluorescent mouse model. A red fluorescent protein-expressing xenograft model

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service