Skip to Content
Merck
All Photos(1)

Key Documents

SMB00579

Sigma-Aldrich

Tacalcitol

≥99% (HPLC)

Synonym(s):

(1α,3β,5Z,7E,24R)-9,10-secocholesta-5,7,10(19)-triene-1,3,24-triol, 1α,24R-dihydroxycholecalciferol, 1α,24R-dihydroxyvitamin D3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

C[C@]1(CCC/2)[C@](CC[C@]1([H])[C@H](C)CC[C@@H](O)C(C)C)([H])C2=C\C=C3C[C@@H](O)C[C@H](O)C\3=C

InChI

1S/C27H44O3/c1-17(2)25(29)13-8-18(3)23-11-12-24-20(7-6-14-27(23,24)5)9-10-21-15-22(28)16-26(30)19(21)4/h9-10,17-18,22-26,28-30H,4,6-8,11-16H2,1-3,5H3/b20-9+,21-10-/t18-,22-,23-,24+,25-,26+,27-/m1/s1

InChI key

BJYLYJCXYAMOFT-RSFVBTMBSA-N

General description

Tacalcitol is a synthetic vitamin D3 analog.[1] It is a vitamin D3 derivative, also known as 1α,24-dihydroxyvitamin D3.[2]

Biochem/physiol Actions

Tacalcitol is widely used in the treatment of dermatosis due to its ability to inhibit keratinocyte proliferation and induce cell differentiation. It also has anti-inflammatory activity and has been shown to reduce cutaneous inflammation via suppression of leukocyte infiltration in skin.[3] Tacalcitol is topically used for the treatment of psoriasis and has also been found to be effective in the treatment of acquired perforating collagenosis (APC), a rare perforating dermatosis.[1] Studies suggest that this compound has potential for the treatment of the hereditary disorder Hailey-Hailey Disease.[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tacalcitol in the treatment of acquired perforating collagenosis.
Escribano-Stable JC
Case Reports in Dermatology, 6(1), 69-73 (2014)
Hailey-hailey disease treated with topical tacalcitol.
Jeon SY et al
Annals of Dermatology, 25(3), 389-390 (2013)
M Fukuoka et al.
British journal of pharmacology, 124(7), 1433-1438 (1998-09-02)
1. The production of chemokines, RANTES and IL-8 in cultured human dermal fibroblasts and the effects of tacalcitol (1alpha,24(R)-dihydroxyvitamin D3) were studied using an enzyme-linked immunosorbent assay. 2. In the unstimulated condition, RANTES and IL-8 were at a trace level

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service