Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H16N5O7S · Na
Molecular Weight:
445.38
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Form:
solid
form
solid
color
off-white
solubility
DMSO: soluble, H2O: soluble
storage temp.
−20°C
Biochem/physiol Actions
A1 adenosine receptor agonist.
Legal Information
Sold under license from the National Institutes of Health.
Disclaimer
Hygroscopic. Photosensitive
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.