Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41103909
Manufacturer/tradename:
Eppendorf® 022665821 (NA), Eppendorf® 5452727007 (EU)
Compatibility:
for use with centrifuge models MiniSpin/MiniSpin Plus
Parameter:
14,100 × g max. RCF
Feature:
lid, to hold 2 x 8-tube PCR strips, fixed angle rotor, non aerosol tight
Product RF55165-EP is not currently sold in your country. Contact Technical Service
feature
lid, to hold 2 x 8-tube PCR strips, fixed angle rotor, non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022665821 (NA), Eppendorf® 5452727007 (EU)
parameter
14,100 × g max. RCF
technique(s)
PCR: suitable, centrifugation: suitable
compatibility
for use with centrifuge models MiniSpin/MiniSpin Plus
General description
Rotors for use with Eppendorf MiniSpin and MiniSpin Plus. Rotors are fixed angle and include quick-Lock lids.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Minispin is a registered trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Rotor for Eppendorf® MiniSpin® / MiniSpin® Plus
Choose from one of the most recent versions:
Already Own This Product?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EP022665821-1EA | 04061835087365 |