Skip to Content
Merck

Skip To

RAB0518

Cell-Based ELISA for detecting Phosphotyrosine

in cultured cell line, 96 assays

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
CZK 10,200.00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
human, mouse, rat
Storage temp.:
−20°C
Shipped in:
wet ice

CZK 10,200.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

96 assays

species reactivity

human, mouse, rat

technique(s)

ELISA: suitable, capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

General description

The Cell-Based Phosphotyrosine ELISA kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cells. It can be used for measuring the relative amount of Phosphotyrosin and screen the effects of various treatments, inhibitors (such as siRNA or chemicals), or activators in cultured human, mouse and rat cell lines.

Immunogen

KLH-conjugated phosphotyramine

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0350RAB0452RAB0448
species reactivity

human, mouse, rat

species reactivity

rat, human, mouse

species reactivity

human, mouse

species reactivity

mouse, human

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

usage

96 assays

usage

96 assays

usage

96 assays

usage

96 assays

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • 5x Blocking Solution (Item F)
    SDS

  • Fixing Solution for Phosphotyrosine Assay (Item D)
    SDS

  • Uncoated Microplate
    SDS

  • Quenching Solution for Cell-based ELISA Assay (Item E)
    SDS

  • Stop Solution
    SDS

  • TMB Substrate Reagent (Item J)
    SDS

  • HRP-Conjugated Phosphotyrosine-specific Antibody Concentrate (Item E)
    SDS

See All (9)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters