Skip to Content
Merck
All Photos(5)

Key Documents

HPA017006

Sigma-Aldrich

Anti-AFM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Afamin precursor, Anti-Alpha-Alb, Anti-Alpha-albumin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:2500- 1:5000

immunogen sequence

INSNKDDRPKDLSLREGKFTDSENVCQERDADPDTFFAKFTFEYSRRHPDLSIPELLRIVQIYKDLLRNCCNTENPPGCYRYAEDKFNETTEKSLKMVQQECKHFQNLGKDGLKYHYLIRLTKIAPQLSTEEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFM(173)

General description

AFM (afamin) is a 75kDa vitamin E-binding glycoprotein belonging to the albumin gene family. It consists of four or five potential N-glycosylation sites. In humans, it is synthesized by cerebrovascular endothelial cells and expressed in human plasma.

Immunogen

Afamin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AFM (afamin) possesses specific binding affinity for the two forms of vitamin E, α-tocopherol and γ-tocopherol. It carries both the forms of vitamin E through the body fluids such as human plasma and follicular fluid under certain conditions, which contributes in the vitamin E transport. It has been reported that AFM may play a role in the fertility and neuro-protection. Increased level of AFM causes insulin resistance and metabolic disorder, polycystic ovary syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86798

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata Seeber et al.
Reproductive biology and endocrinology : RB&E, 12, 88-88 (2014-09-12)
High plasma concentrations of the vitamin E-binding protein afamin have been previously shown to be associated with insulin resistance and metabolic syndrome. We set out to determine whether the concentration of afamin in the serum of women with polycystic ovarian
Andreas F Voegele et al.
Biochemistry, 41(49), 14532-14538 (2002-12-05)
Human plasma afamin, the fourth member of the albumin gene family, is shown to be a specific binding protein for vitamin E. A radio ligand-binding assay followed by Scatchard and Hill analysis are used to show that afamin has a
Lidija Jerkovic et al.
Journal of proteome research, 4(3), 889-899 (2005-06-15)
Hydrophobic vitamins are transported in human plasma and extravascular fluids by carrier proteins. No specific protein has been described so far for vitamin E, which plays a crucial role in protecting against oxidative damage and disease. We report here the
Carsten Behr Andersen et al.
Arteriosclerosis, thrombosis, and vascular biology, 38(9), 2254-2267 (2018-07-14)
Objective- Porosity of the intraluminal thrombus (ILT) is believed to convey biologically active components from the bloodstream toward the aneurismal wall. Accumulation of molecules in the abdominal aortic aneurysmatic tissue may influence vascular protein turnover and regulate abdominal aortic aneurysm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service