Skip to Content
MilliporeSigma

E5149

EMB broth

pH 7.2±0.2 (25 °C)

Synonym(s):

Eosin Methylene Blue Broth

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171606
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


pH

7.2±0.2 (25 °C)

Quality Segment

Application

Recommended for the isolation and differentiation of gram negative enteric bacteria.

Other Notes

Components (g/L):
Peptic Digest of Animal Tissue 10.00
Dipotassium Phosphate 2.00
Lactose 5.00
Sucrose 5.00
Eosin-Y 0.40
Methylene Blue 0.065




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library