Skip to Content
Merck
All Photos(1)

Documents

B3402

Sigma-Aldrich

Ethyl 6-bromohexanoate

≥97% (GC)

Synonym(s):

6-Bromohexanoic acid ethyl ester, Ethyl 6-bromocapronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)5COOCH2CH3
CAS Number:
Molecular Weight:
223.11
Beilstein:
1756051
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (GC)

refractive index

n20/D 1.458 (lit.)

bp

128-130 °C/16 mmHg (lit.)

density

1.254 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCCBr

InChI

1S/C8H15BrO2/c1-2-11-8(10)6-4-3-5-7-9/h2-7H2,1H3

InChI key

DXBULVYHTICWKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.9 °F - closed cup

Flash Point(C)

58.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service