sterility
sterile; γ-irradiated
packaging
pkg of 1
capacity
20 L
volume
20 L , 20000 mL
General description
Configured with:
Port P1: 1 x 2.5 ft. length of Size 15 Opaque 001 C-Flex® Tubing with 1/4" Male Quick Connect with Cap
Port P2: 1 x 2.5 ft. length of Size 15 Opaque 001 C-Flex® Tubing with 1/4" Female Quick Connect with Cap
Port P3: 1 Spike Port with short tubing extension
Features and Benefits
BIOEAZE Ethyl Vinyl Acetate (EVA) bags are constructed from a single-web, medical grade laminate of four different films, with a fluid contact surface of EVA, with two layers of low-density polyethylene (LDPE) enveloping a layer of ethyl vinyl alcohol (EVOH).
Packaging
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility γ-irradiated | sterility - |
| capacity 20 L | capacity 1 L | capacity 5 L | capacity - |
| packaging pkg of 1 | packaging pkg of 5 | packaging pkg of 5 bags | packaging pkg of 25 pkg |
| volume 20 L | volume 1 L , 1000 mL | volume 5 L , 5000 mL | volume - |
Still not finding the right product?
Explore all of our products under BIOEAZE™ Ethyl Vinyl Acetate (EVA) Bags
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
BIOEAZE™ bags and disposable bioprocessing systems are designed by SAFC Biosciences to address your specific needs and applications. Disposable technology provides many advantages, including the elimination of cleaning and sterilization procedures and the validation of these processes. BIOEAZE disposable bioprocess bags are routinely used as flexible containers for the pharmaceutical and biotechnology industries. BIOEAZE bioprocess bags are specifically designed to provide strong seals, extraordinary robustness, superior flex, crack and pin-hole resistance, plus desirable gas and moisture barrier performance.
BIOEAZE™ Polyethylene (PE) and Ethyl Vinyl Acetate (EVA) bioprocess bags are designed to hold a variety of sterile liquids including cell culture media, water and buffers. SAFC Biosciences has performed extensive physical and chemical characterizations of these films, including Total Organic Carbon (TOC) testing.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1330B | 04061834411079 |
