Skip to Content
Merck
All Photos(1)

Documents

BCR097

Benzo[a]fluoranthene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1957846
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc-2c(c1)-c3cccc4cc5ccccc5c-2c34

InChI

1S/C20H12/c1-2-8-15-13(6-1)12-14-7-5-11-17-16-9-3-4-10-18(16)20(15)19(14)17/h1-12H

InChI key

OQDXASJSCOTNQS-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR097

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E H Weyand et al.
Cancer letters, 52(3), 229-233 (1990-07-31)
The tumor-initiating activities of benzo[a]fluoranthene (BaF), benzo[b]fluoranthene (BbF), naphtho[1,2-b]fluoranthene (NbF) and naphtho[2,1-a]fluoranthene (NaF) were evaluated on the skin of female CD-1 mice. Each of these polycyclic aromatic hydrocarbons was assayed at total initiation doses of 1.0 and 4.0 mumol/mouse. These
Han Yeong Kaw et al.
The Analyst, 143(19), 4575-4584 (2018-08-17)
In order to extract trace organic compounds (TrOCs) from large volumes of water (over 10 L), an ultrasound-assisted liquid-liquid spray extraction (UA-LLSE) technique is proposed. The UA-LLSE integrates liquid-liquid extraction, spray atomisation and ultrasonication in a single step. Under an
Gabriel A Odugbesi et al.
Journal of chromatography. A, 1604, 460466-460466 (2019-09-03)
Ionic liquids (ILs) are well-known in the field of separation science for their unique selectivity when used as stationary phases in gas chromatography (GC). While a significant amount of knowledge has been attained in correlating structural features of an IL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service