Skip to Content
Merck
All Photos(1)

Documents

ARK2064

SAFC

PROTHIPENDYLHYDROCHLORIHYDRAT (DOMINAL HCL)

Pharma Manufacturing

Synonym(s):

Dominal hydrochloride, 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H19N3HCLH2O
UNSPSC Code:
12352200

mol wt

339.91 g/mol

Quality Level

mp

108-112 °C (lit.)

storage temp.

room temp

InChI

1S/C16H19N3S.ClH/c1-18(2)11-6-12-19-13-7-3-4-8-14(13)20-15-9-5-10-17-16(15)19;/h3-5,7-10H,6,11-12H2,1-2H3;1H

InChI key

CQJSAKJMCVSEGU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service