Skip to Content
Merck

Skip To

93085

Abberior® STAR 580, biotin

≥90.0% (HPCE)

Sign In to View Organizational & Contract Pricing.



assay

≥90.0% (HPCE)

form

liquid

manufacturer/tradename

Abberior GmbH

fluorescence

λex 587 nm; λem 607 nm±10 nm in DMSO

detection method

fluorometric

storage temp.

2-8°C

General description

Abberior® STAR 580 dye was developed for STED and confocal microscopy in the orange spectral region. The product, ”Abberior” STAR 580, biotin is a derivative of Abberior® STAR 580 type of dyes. It can be effectively excited with an excitation wavelength between 500 - 600 nm. For STED microscopy, it can be used most efficiently with a STED laser wavelength between 750 - 800 nm. Abberior® STAR 580 dyes can substitute dyes like Cy3, ATTO 590 or Abberior® 568. These are ideal partners for abberior STAR RED or abberior STAR 635P to obtain optimal 2 color STED results.

Application

Designed and tested for fluorescent super-resolution microscopy

Features and Benefits

  • Fluorescent dye for physical/chemical research purposes, spectroscopy, and optical microscopy in biological and biomedical applications.
  • High photostability.
  • High fluorescent quantum yield of 90%.
  • Suitable for fluorescence labeling of avidin- or streptavidin-modified molecules.

Legal Information

Atto is a trademark of Atto-Tec GmbH
abberior is a registered trademark of Abberior GmbH


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
93085-1MG04061835503766

Questions

Reviews

No rating value

Active Filters