Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 25 mg | Please contact Customer Service for Availability | CZK 7,670.00 |
About This Item
grade
analytical standard
Quality Segment
assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
OCCN(CCO)C(C=C1C)=CC=C1/N=N/C2=CC=C([N+]([O-])=O)C=C2
InChI
1S/C17H20N4O4/c1-13-12-16(20(8-10-22)9-11-23)6-7-17(13)19-18-14-2-4-15(5-3-14)21(24)25/h2-7,12,22-23H,8-11H2,1H3/b19-18+
InChI key
ADCWDMYESTYBBN-VHEBQXMUSA-N
General description
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable |
| format neat | format neat | format neat | format neat |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| grade analytical standard | grade analytical standard | grade analytical standard | grade analytical standard |
| assay ≥98.0% (HPLC) | assay ≥96.0% (HPLC) | assay ≥96.0% (HPLC) | assay ≥96.0% (HPLC) |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life - | shelf life limited shelf life, expiry date on the label |
signalword
Warning
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1B
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 79892-25MG | 04061832973920 |




