Skip to Content
Merck
All Photos(1)

Documents

238562

Sigma-Aldrich

IGEPAL® CA-210

average Mn ~294

Synonym(s):

Polyoxyethylene (2) isooctylphenyl ether, Polyoxyethylene (2) octylphenyl ether, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)n · C14H22O n~1.5
CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

mol wt

average Mn ~294

refractive index

n20/D 1.509 (lit.)

density

1.009 g/mL at 25 °C (lit.)

HLB

4

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Legal Information

IGEPAL is a registered trademark of Solvay

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell M Morphew et al.
Veterinary parasitology, 191(3-4), 379-385 (2012-10-02)
Fasciola hepatica NP-40 released protein extract (FhNPE) exhibits potent Th1 immunosuppressive properties in vitro and in vivo. However, the protein composition of this active fraction, responsible for Th1 immune modulatory activity, has yet to be resolved. Therefore, FhNPE, a Nonidet
Rui Ma et al.
Analytical biochemistry, 424(2), 124-126 (2012-03-01)
Triton X-114 temperature transition extraction has been considered to be a simple and cost-effective strategy to eliminate endotoxin from plasmid preparations. However, a repeated cooling-heating process may promote the degradation of plasmid DNA. Based on the finding that the cloud
Zonghao Li et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(2), 247-253 (2012-11-22)
A new, simple and rapid method based on dispersive liquid-liquid microextraction (DLLME) was developed for extracting and preconcentrating copper (Cu), nickel (Ni), lead (Pb) and cadmium (Cd) in water samples prior to flame atomic absorption spectrometry (FAAS) analysis. 1-(2-thiazolylazo)-naphthol (TAN)
D V Krishna Pantakani et al.
PloS one, 6(12), e29584-e29584 (2012-01-05)
ZFYVE27 (Protrudin) was originally identified as an interacting partner of spastin, which is most frequently mutated in hereditary spastic paraplegia. ZFYVE27 is a novel member of FYVE family, which is implicated in the formation of neurite extensions by promoting directional
Halil İbrahim Ulusoy et al.
Talanta, 88, 516-523 (2012-01-24)
A new micelle-mediated separation and preconcentration method was developed for ultra-trace quantities of mercury ions prior to spectrophotometric determination. The method is based on cloud point extraction (CPE) of Hg(II) ions with polyethylene glycol tert-octylphenyl ether (Triton X-114) in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service