RIE156320647
Rieke Metals, Inc.
4-Acetoxy-2'-chlorobenzophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-(2-chlorobenzoyl)phenyl acetate
Inchi Code
1S/C15H11ClO3/c1-10(17)19-12-8-6-11(7-9-12)15(18)13-4-2-3-5-14(13)16/h2-9H,1H3
InChI key
KWEFKRBEISXAIJ-UHFFFAOYSA-N
Purity
90%
Country of Origin
US
Product Link
https://riekemetals.com/products-services/catnum/5177-14
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service