Recommended Products
biological source
human
Quality Level
packaging
vial of >1X10⁶ cells
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell culture | mammalian: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
Application
- Each vial contains > 1X106 viable cells.
- Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
- Cells are verified to be of mouse origin and negative for inter-species contamination from rat, human, chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
- Cells are negative for mycoplasma contamination.
The great promise of immune-based therapies in cancer and recent progress in successful application of these approaches has brought to the fore the necessity of immune-competent models to evaluate immune system responses to cancer cells. Melanomas exhibit relatively high somatic mutation burden, and these mutations may act as neoantigens that generate anti-tumor immune responses. The development of immunocompetent cell models is critical to the advancement of cancer immunotherapy and understanding of immunity, although few tractable model systems are available.
References
1. Pigment Cell Melanoma Res 2016, 29(5): 590-597.
2. Nat Genet 2009, 41(5): 544-552.
3. Pigment Cell Melanoma Res 2017, 30(4): 428-435.
4. Ramseier JY, Charos A, Park K, Damsky W, Bosenberg MW (2019). Cancer Res 79(13 Suppl) Abstract #4622.
References
1. Pigment Cell Melanoma Res 2016, 29(5): 590-597.
2. Nat Genet 2009, 41(5): 544-552.
3. Pigment Cell Melanoma Res 2017, 30(4): 428-435.
4. Ramseier JY, Charos A, Park K, Damsky W, Bosenberg MW (2019). Cancer Res 79(13 Suppl) Abstract #4622.
Features and Benefits
The characteristics of the YUMMER1.G1F cell line make it a valuable model for investigating sex-related differences in immune checkpoint inhibition and mechanisms of anti-tumor responses.
Storage and Stability
The cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting functionality.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.;<bold>This product contains genetically modified organisms (GMO). </bold>Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges to request certain information about you and the establishment where the GMOs are being handled. <IS7_ContentPageCategoryLink target="Up.b.qB.CN8AAAFAnd8QWRbW">Click here for Enduser Declaration (EUD) Form</IS7_ContentPageCategoryLink>.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service