Skip to Content
Merck

Skip To

MABS830

Anti-Acetyl CoA Carboxylase 1 Antibody, clone 7H4.2

clone 7H4.2, from mouse

Synonym(s):

ACC1, ACC-alpha

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,040.00
100 μg

Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7H4.2, monoclonal
Application:
IHC, WB
Citations:
-

CZK 4,040.00


Available to ship TODAYDetails

A recombinant, preservative-free antibody is available for your target. Try ZRB1484

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7H4.2, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ACACA(31)
mouse ... Acaca(107476)

General description

Acetyl-CoA carboxylase 1 (UniProt: Q13085; also known as ACC1, ACC-alpha) is encoded by the ACACA (also known as ACAC, ACC1, ACCA) gene (Gene ID: 31) in human. It catalyzes the rate-limiting reaction in the biogenesis of long-chain fatty acids. ACC1 is a multi-subunit, biotin-dependent enzyme that catalyzes carboxylation of acetyl-CoA to produce malonyl-CoA through its two catalytic activities, biotin carboxylase (BC) and carboxyltransferase (CT). The activity of ACC can be controlled at the transcriptional level as well as by small molecule modulators and covalent modification. Two isoforms fof ACC have been reported, alpha and beta that are encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. ACC1 levels are regulated under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. ACC1 activity is inhibited upon phosphorylation, which occurs when glucagon or epinephrine binds to the receptors or when the energy status of the cell is low, leading to the activation of the AMP-activated protein kinase. The presence of fatty acids inhibits the activities of ACC1. ACC is considered as an important target in the design of new anti-obesity drugs.
~265 kDa observed; 265.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST/His-tagged recombinant fragment corresponding to 86 amino acids from the C-terminal end of human Acetyl-CoA carboxylase 1.

Application

Anti-Acetyl CoA Carboxylase 1 Antibody, clone 7H4.2, Cat. No. MABS830, is a mouse monoclonal antibody that targets Acetyl CoA Carboxylase 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Acetyl CoA Carboxylase 1 in human tonsil, human testis, and human cervical cancer tissue.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Acetyl CoA Carboxylase 1 in 10 µg of NIH/3T3 cell lyate.

Biochem/physiol Actions

Clone 7H4.2 detects acetyl-CoA carboxylase 1 in human. It targets an epitope within 86 amino acids from the C-terminal end.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Acetyl CoA Carboxylase 1 in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABS2024ABS1068SAB4501395
clone

7H4.2, monoclonal

clone

10H2.1, monoclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, human

species reactivity

human

species reactivity

human

species reactivity

mouse, rat, human

Gene Information

human ... ACACA(31)
mouse ... Acaca(107476)

Gene Information

human ... ACACA(31)

Gene Information

human ... ACACA(31)

Gene Information

human ... ACACA(31)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters