Skip to Content
Merck
All Photos(1)

Documents

MABF3091

Sigma-Aldrich

Anti-LMP1 Antibody, clone 1G6

new

Synonym(s):

LMP-1, Protein p25, Protein p63

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G6, monoclonal

mol wt

calculated mol wt 42 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2a

epitope sequence

C-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

vaccinia virus ... LMP1(3783750)

Specificity

Clone 1G6 is a rat monoclonal antibody that detects LMP1. It targets an epitope within the C-terminal cytoplasmic domain of the protein.

Immunogen

Wild type LMP1-TrpE-fusion protein from Epstein-Barr virus.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in B95-8 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected LMP1 in B95-8 cells.

Tested Applications

Immunohistochemistry Analysis: A representative lot detected LMP1 in Immunohistochemistry applications (Nicholls, J., et al. (2004). J Virol Methods. 116(1):79-88; Shair, K.H.Y., et al. (2007). PLoS Pathog. 3(11):e166).

Immunocytochemistry Analysis: A representative lot detected LMP1 in Immunocytochemistry applications (Nicholls, J., et al. (2004). J Virol Methods. 116(1):79-88).

Western Blotting Analysis: A representative lot detected LMP1 in Western Blotting applications (Nicholls, J., et al. (2004). J Virol Methods. 116(1):79-88; Thornburg, N.J., et al. (2006). Oncogene. 25(2):288-97; Hannigan, A., et al. (2010). Mol Cancer. 9:9:184; Rancan, C., et al. (2015). PLoS Pathog. 11(6):e1004906).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Latent membrane protein 1 (LMP1) (UniProt: P03230) is encoded by the LMP1 (also known as BNLF1) gene (Gene ID: 3783750) in Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4). Latent membrane protein 1 (LMP1) is a viral oncoprotein encoded by the Epstein-Barr virus (EBV) and is known for its critical role in EBV-associated oncogenesis. LMP1 functions as a constitutively active mimic of the tumor necrosis factor receptor (TNFR) and can activate various signaling pathways, including NF- B, JNK, and PI3K/Akt. Through its ability to modulate these pathways, LMP1 can promote cell proliferation, inhibit apoptosis, and induce the expression of various cytokines and growth factors. Additionally, LMP1 contributes to the immortalization of infected B cells and is implicated in the pathogenesis of EBV-associated malignancies, including Hodgkin′s lymphoma and nasopharyngeal carcinoma.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service