Skip to Content
MilliporeSigma

MABF2008

Anti-IL-17F Antibody, clone MM17F-8F5

Synonym(s):

Interleukin-17F

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
MM17F-8F5, monoclonal
Application:
ELISA, FACS, Neutral, WB, inhibition assay
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MM17F-8F5, monoclonal

mol wt

calculated mol wt 17.94 kDa, observed mol wt ~15 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable, flow cytometry: suitable, inhibition assay: suitable, neutralization: suitable, western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Il17f(257630)

General description

Interleukin-17F (UniProt: Q7TNI7; also known as IL-17F) is encoded by the Il17f gene (Gene ID: 257630) in murine species. The IL-17 family of cytokines contains six members that share four conserved cysteine residues at their C-terminal region that participate in the formation of intermolecular disulfide linkages. Members of this family are associated with the pathogenesis of multiple autoimmune diseases including rheumatoid arthritis, multiple sclerosis, and inflammatory bowel diseases. IL-17F is an effector cytokine of innate and adaptive immune system involved in antimicrobial host defense and maintenance of tissue integrity. It is a disulfide-linked homodimeric protein that is synthesized with a signal peptide (aa 1-28), which is subsequently cleaved off to produce mature secreted protein (aa 29-161). It can also form a heterodimer with IL-17A. Its actions are mediated via interaction with IL-17RA and IL-17RC receptors with 2:1 binding stoichiometry. It is primarily involved in host defense against extracellular bacteria and fungi by inducing neutrophilic inflammation. As signature effector cytokine of T-helper 17 cells (Th17), it induces neutrophil activation and recruitment at infection and inflammatory sites. It is also known to stimulate the production of antimicrobial β-defensins by mucosal epithelial cells thereby limiting the entry of microbes through the epithelial barriers. (Ref.: St. Leger, AJ., et al. (2017). Immunity 47(1); 148 158; Chang, SH., and Dong, C. (2009). Cytokine.46(1); 7-11).

Immunogen

Ovalbumin-conjugated, full-length recombinant murine IL-17F.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant mouse IL-17F.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant mouse IL-17F.

Tested Applications

Flow Cytometry Analysis: A representative lot detected Interleukin-17F in Flow Cytometry application (St. Leger, A.J., et al. (2017). Immunity. 47(1):148-158.e5).

ELISA Analysis: Various dilutions of this antibody detected recombinant mouse IL-17F.

Neutralizing: A representative lot neutralized IL-17F activity and reduced neutrophil chemotactic activity produced by IL-17A / splenocytes activated under Th17 conditions.(Uyttenhove, C., et al. (2011). J Leukoc Biol. 89(6):1001-7).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone MM17F-8F5 is a mouse monoclonal antibody that detects Interleukin-17F.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library