Skip to Content
Merck
All Photos(2)

Documents

MABE1057

Sigma-Aldrich

Anti-ADA3 Antibody, clone 5C9/C8

clone 5C9/C8, from mouse

Synonym(s):

Transcriptional adapter 3, ADA3 homolog, ADA3-like protein, Alteration/deficiency in activation 3, hADA3, STAF54, Transcriptional adapter 3-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C9/C8, monoclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TADA3(10474)

General description

Transcriptional adapter 3 (UniProt O75528; also known as ADA3 homolog, ADA3-like protein, Alteration/deficiency in activation 3, hADA3, STAF54, Transcriptional adapter 3-like) is encoded by the TADA3 (also known as ADA3, TADA3L) gene (Gene ID 10474) in human. Alteration/deficiency in activation 3 (ADA3) is a component of several transcriptional co-activator and histone acetyltransferase (HAT) complexes and plays a critical role in in cell cycle regulation. Ada3 deletion in mice is embryonic lethal, and Ada3-deficient mouse embryonic fibroblasts (MEFs) exhiit a severe proliferation defect, dramatic changes in global histone acetylation, mitotic defects, as well as a delay in G2/M-to-G1 and G1-to-S transition. ADA3 also plays a role in genomic stability by controlling DNA repair checkpoints. ChIP-seq analsis reveals that ADA3 is significantly associated with human centromere regions across most chromosomes. In addition, ADA3 is found associated with CENP-B throughout all phases of the cell cycle, and CENP-B centromere binding decreased upon ADA3 knockdown. Wild-type human ADA3, but not CENP-B-binding deficient ADA3 mutant, prevented cell proliferation defects in MEFs following endogenous mouse ADA3 knockown. ADA3 overexpression and mislocalization correlates with poor prognosis in breast cancer patients.

Specificity

Clone 5C9/C8 specifically detected Cre recombinase expression-induced ADA3 downregulation in Ada3FL/FL MEFs. Clone 5C9/C8 immunostained the nuclei of untransfected, but not ADA3 shRNA-transfected, 76N-TERT human mammary epithelial cells (Mohibi, S., et al. (2015). J. Biol. Chem. 290(47):28299-28310; Mohibi, S., et al. (2012). J. Biol. Chem. 287(35):29442-29456).

Immunogen

Full-length recombinant human ADA3.

Application

Anti-ADA3, clone 5C9/C8, Cat. No. MABE1057, is a highly specific mouse monoclonal antibody that targets TADA3 and has been tested in Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected nuclear ADA3 immunoreactivity in formalin-fixed, paraffin-embedded human breast carcinoma tissue section (Courtesy of Vimla Band, Ph.D., University of Nebraska USA).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected ADA3 occupancy at the X chromosome centromere HOR region, the kinetochore assembly site also occupied by CENP-A and CENP-B, using TERT-immortalized human mammary epithelial cell 76N-TERT chromatin preparation. shRNA-mediated ADA3 knockdown led to a reduction in CENP-B, but not CENP-A, recruitment to the HOR region (Mohibi, S., et al. (2015). J. Biol. Chem. 290(47):28299-28310).

Immunocytochemistry Analysis: A representative lot detected ADA3 nuclear immunoreactivity in untransfected, but not ADA3 shRNA-transfected, TERT-immortalized human mammary epithelial 76N-TERT cells by fluorescent immunocytochemistry staining of 4% formaldehyde-fixed cells. Dual fluorescent staining revealed ADA3 and CENP-B nuclear colocalization (Mohibi, S., et al. (2015). J. Biol. Chem. 290(47):28299-28310).

Western Blotting Analysis: Representative lots detected both the endogenous mouse ADA3 (mADA3) and the exogenously expressed human ADA3 (hADA3) in Ada3FL/FL MEFs virally infected to express FLAG-tagged full-length or a.a. 111-432 hADA3 constructs. Cre recombinase expression downregulated only mADA3, but not hADA3 (Mohibi, S., et al. (2015). J. Biol. Chem. 290(47):28299-28310; Mohibi, S., et al. (2012). J. Biol. Chem. 287(35):29442-29456).

Western Blotting Analysis: A representative lot detected ADA3 expression levels among a panel of mouse tissues (Mohibi, S., et al. (2012). J. Biol. Chem. 287(35):29442-29456).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ADA3 in 10 µg of MCF-7 cell lysate.

Target description

~55 kDa observed. 48.90 kDa (human and mouse isoform 1) calculated. The larger-than-calculated band size is consistent with that reported in the literature (Mohibi, S., et al. (2015). J. Biol. Chem. 290(47):28299-28310). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service