Skip to Content
Merck

Skip To

MAB8043

Anti-Adenovirus 1,2,5,6 Antibody, clone 7/48.7c

ascites fluid, clone 7/48.7c, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 12,100.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
7/48.7c, monoclonal
Species reactivity:
human
Application:
IF
Citations:
4

CZK 12,100.00


Estimated to ship onJune 03, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

ascites fluid

clone

7/48.7c, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1

shipped in

wet ice

General description

Adenoviruses are 65-80 nm, non-enveloped, regular icosahedron pathogens often associated with respiratory and gastrointestinal illness as well as conjunctivitis. Well over 40 types of adenoviruses have currently been recognized. In addition to their deleterious effects, adenoviruses have been used in vaccine production and in gene therapy. They also have the ability to trigger cell proliferation by interfering with host cellular anti-oncogenes. Since they are easy to prepare, multiple quickly and efficiently, and have the ability to infect and deliver their genome (including researcher-created constructs) to the nucleus of a variety of post-mitotic cells, adenoviruses are extremely valuable in research and medical applications ranging from virus/host cell interaction to the insertion of novel genes or regulatory mechanisms into eukaryotic cellular systems, to delivery of therapeutic anti-cancer genes.

Application

Anti-Adenovirus 1, 2, 5, 6 Antibody, clone 7/48.7c detects level of Adenovirus 1 & has been published & validated for use in IF.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB8051MAB805MAB8052
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

clone

7/48.7c, monoclonal

clone

2/6, monoclonal

clone

2/6, monoclonal, 20/11, monoclonal

clone

20/11, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

ascites fluid

antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

purified immunoglobulin

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

technique(s)

immunofluorescence: suitable

technique(s)

ELISA: suitable, flow cytometry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

ELISA: suitable, flow cytometry: suitable, immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
MAB804304053252357091

Questions

Reviews

No rating value

Active Filters