Skip to Content
Merck

Skip To

ABC1687

Anti-Heparanase-1

from rabbit, purified by affinity chromatography

Synonym(s):

Endo-glucoronidase, Heparanase, Hpa1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... HPSE(10855)

General description

Heparanase (UniProt: Q9Y251; also known as EC: 3.2.1.166, Endo-glucuronidase, Heparanase-1, Hpa1) is encoded by the HPSE (also known as HEP, HPA, HPA1, HPR1, HPSE1, HSE1) gene (Gene ID: 10855) in human. Heparanase is a heterodimeric, glycosylated, endoglycosidase that cleaves heparan sulfate proteoglycans (HSPGs) into heparan sulfate side chains and core proteoglycans and participates in extracellular matrix (ECM) degradation and remodeling. It is known to selectively cleave the linkage between a glucuronic acid unit and an N-sulfo glucosamine unit carrying either a 3-O-sulfo or a 6-O-sulfo group. It is also shown to cleave the linkage between a glucuronic acid unit and an N-sulfo glucosamine unit carrying a 2-O-sulfo group, but not linkages between a glucuronic acid unit and a 2-O-sulfated iduronic acid moiety. Heparanase is highly expressed in placenta and spleen and weakly expressed in lymph node, thymus, peripheral blood leukocytes, bone marrow, endothelial cells, fetal liver, and tumor tissues. Heparanase is synthesized with a signal peptide (aa 1-35) and a propeptide chain (110-157), which are cleaved off to produce the heterodimeric mature form of 8 kDa and 50 kDa subunits. This active 8/50 kDa heterodimer is resistant to degradation. Heparanase is essentially inactive at neutral pH but becomes active under acidic conditions such as during tumor invasion and in inflammatory processes. Heparanase activity is inhibited by EDTA and laminarian sulfate. Four isoforms of heparanase have been described that are produced by alternative splicing.
~59 kDa observed; 61.15 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal half of larger subunit (50 kDa)of human Heparanase-1.

Application

Anti-Heparanase-1, Cat. No. ABC1687, is a rabbit polyclonal antibody that detects Heparanase-1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Heparanase-1 in human spleen and human placenta tissue sections.

Western Blotting Analysis: 1 µg/mL from a representative lot detected Heparanase-1 in human spleen tissue lysate.
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

This rabbit polyclonal antibody detects human Heparanase-1. It targets an epitope with in 13 amino acids from the C-terminal half of larger subunit (50 kDa).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Heparanase-1 in NIH/3T3 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB1948PABC947MAB2600
biological source

rabbit

biological source

rat

biological source

rabbit

biological source

mouse

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... HPSE(10855)

Gene Information

human ... HSPG2(3339)

Gene Information

human ... SH3GLB1(51100)

Gene Information

human ... GPC1(2817)
rat ... Gpc1(58920)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

species reactivity

mouse, human

species reactivity

human

species reactivity

human

species reactivity

human, rat


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABC1687-100UG04054839623462
ABC1687-25UG04054839623479

Questions

Reviews

No rating value

Active Filters