Skip to Content
MilliporeSigma

8.55129

NovaSyn® TG PAP RESIN

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

NovaSyn® TG, Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

NovaSyn TG PAP resin contains an acid cleavable linkage between the polystyrene matrix and the PEG chain. Treatment of peptides prepared on this resin with TFA/thioanisole (95:5) for 12 hours or TMSBr/TFA/thioanisole (1:94:5) for 60 minutes releases a fully deprotected peptide possessing a PEG-chain attached to the C-terminus [1]. Such peptide-PEG conjugates can be used directly as immunogens, without the need for a carrier protein. The peptide-PEG conjugates are also highly soluble and provide an effective solution to insoluble peptides.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Literature references

[1] W. Rapp in ′Combinatorial Peptide and Nonpeptide Libraries′, G. Jung (Eds), Vch, 1996, pp. 425.

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 0.30 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library