Skip to Content
Merck

Skip To

79158BC

GDS PPMX Surface Pro Pen

BioControl Systems, for use with GDS PPMX Microsoft Surface Laptop

Synonym(s):

Active Pen, Digital Pen, Digital Stylus, Microsoft Pen, Stylus Pen, Surface Pen, Surface Pro Pen, Tablet Pen, Touch Pen, Wireless Pen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
CZK 3,220.00

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81

CZK 3,220.00


Please contact Customer Service for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

for use with GDS PPMX Microsoft Surface Laptop

storage temp.

room temp

General description

The GDS PickPen® PIPETMAX® (PPMX) Surface Pro Pen is a digital pen designed for use with a GDS PPMX Microsoft Surface Laptop. It offers precise and responsive pen input, allowing users to write, draw, and interact with their Surface Pro device more intuitively. The pen features pressure sensitivity and tilt functionality, providing a natural and realistic writing and drawing experience. It is a popular accessory among digital artists, note-takers, and professionals who use Microsoft Surface Pro laptops/tablets.

Application

The GDS PPMX Surface Pro Pen is to be used with the GDS PPMX Microsoft Surface Laptop for various applications.

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79146BC79147BC79145BC
compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

BioControl GDS

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

-

packaging

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

Assurance GDS® PickPen, BioControl Systems 79145

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters