Skip to Content
Merck
All Photos(1)

Key Documents

540020

Sigma-Aldrich

Pluripotin

Synonym(s):

Pluripotin, Ras GTPase-Activating Protein Inhibitor II, SC1, N-(3-(7-(1,3-dimethyl-1H-pyrazol-5-ylamino)-1-methyl-2-oxo-1,2-dihydropyrimido[4,5-d]pyrimidin-3(4H)-yl)-4-methylphenyl)-3-(trifluoromethyl)benzamide, ERK Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25F3N8O2
Molecular Weight:
550.54
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable dihydropyrimido-pyrimidine compound that effectively upregulates GTP-bound active Ras level and inhibits LIF- (leukemia inhibitory factor) induced Erk1/2 phosphorylation in OG2-mES (murine Embryonic Stem) cells (1 µM) via direct interaction with Erk and RasGAP (KD = 212 and 98 nM in binding RasGAP 729-1042 catalytic domain and Erk1, respectively), without affecting cellular STAT3 or GSK-3β/Wnt signaling. Pluripotin/SC1 is shown to greatly complement and enhance the efficiency of LIF in deriving ESCs from blastocysts of several murine strains and effectively maintain the pluripotent state of the ESCs even after 11 passages in cultures. SC1-expanded ESCs effectively form EBs (embryonic bodies) in vitro upon SC1 and LIF withdrawal and are capable of generating chimeric offsprings from surrogate mice when injected into blastocysts from different strains in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yang, W., et al. 2009. Stem Cells27, 383.
Chen, S., et al. 2006. Proc. Natl. Acad. Sci. USA103, 17266.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service