Skip to Content
Merck
All Photos(1)

Documents

203715

Sigma-Aldrich

BQ-610 - Calbiochem

A highly selective ETA receptor antagonist (IC₅₀ = 20 nM) that attenuates ET-induced reduction in cardiac output.

Synonym(s):

BQ-610 - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H47N7O6
Molecular Weight:
673.80
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

lyophilized powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A highly selective ETA receptor antagonist (IC50 = 20 nM) that attenuates ET-induced reduction in cardiac output. Also inhibits ET-1- and ET-3-induced activation of adenylate cyclase (IC50 = 28.2 nM). BQ-610 is 30-fold more potent than BQ-123 (Cat. No. 05-23-3831) in porcine aortic smooth muscle membrane assay.
A highly selective endothelin A (ETA) receptor antagonist (IC50 = 20 nM) that attenuates ET-induced reduction in cardiac output. Inhibits ET-1 induced mitogenesis in airway smooth muscle cells in a concentration-dependent manner (IC50 = 20 nM). Also inhibits ET-1 and ET-3 induced activation of adenylate cyclase (IC50 = 28.2 nM). Has been shown to prevent endothelin-induced vasoconstriction. BQ-610 is 30-fold more potent than BQ-123 (Cat. No. 05-23-3831) in a porcine aortic smooth muscle assay.

Biochem/physiol Actions

Cell permeable: no
Primary Target
ETA receptor antagonist
Product does not compete with ATP.
Reversible: no
Target IC50: 20 nM as ETA receptor antagonist that attenuates ET-induced reduction in cardiac output

Warning

Toxicity: Standard Handling (A)

Sequence

Homopiperidinyl-CO-Leu-D-Trp(CHO)-D-Trp-ONH4

Physical form

Supplied as an ammonium salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Beyer, M.E., et al. 1999. Hypertension33, 145.
Carratu, P., et al. 1997. Am. J. Physiol.272, L1021.
Illing, B., et al. 1996. J. Cardiovasc. Pharmacol.27, 487.
Han, H., et al. 1995. J. Mol. Cell. Cardiol.27, 761.
Ishikawa, K., et al. 1992. in Peptides, Schneider and Eberle, eds. (ESCOM), p. 685.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service