Skip to Content
Merck
All Photos(2)

Key Documents

06-1038

Sigma-Aldrich

Anti-Unc84B/SUN2 Antibody

0.5 mg/mL, from rabbit

Synonym(s):

Rab5-interacting protein, Sad1 unc-84 domain protein 2, Sad1/unc-84 protein-like 2, nuclear envelope protein, unc-84 homolog B, unc-84 homolog B (C. elegans)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, canine

species reactivity (predicted by homology)

mouse (87%), dog, horse, rat (87%)

concentration

0.5 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SUN2(25777)

General description

UNC84B (Protein UNC-84 homolog B or SUN2), is a single-pass nuclear envelope transmembrane protein responsible for nuclear migrations that are essential for development. UNC-84B has a predicted transmembrane domain and a C-terminal region with similarity to the S. pombe spindle pole body protein Sad1. UNC-84B interacts with RAB5A and is widely expressed, and highly expressed in the heart, lungs and muscles. UNC-84 may function to facilitate a nuclear-centrosomal interaction required for nuclear migration and anchorage.

Specificity

This antibody recognizes Unc84B/SUN2 at the N-terminus.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to human Unc84B/SUN2 at and around the N-terminus.

Application

Immunoprecipitation Analysis: 10 µg from a previous lot immunoprecipitated Unc84B/SUN2 from 500 µg of U2OS cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-Unc84B/SUN2 Antibody is validated for use in Immunoprecipitation and Western Blotting for the detection of Unc84B/SUN2.

Quality

Evaluated by Western Blotting in U2OS cell lysate.

Western Blot (SNAP ID) Analysis: 1:250 dilution of this antibody detected Unc84B/SUN2 on 10 µg of U2OS cell lysate.

Target description

~80 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
U2OS cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayaka Matsumoto et al.
Cancer medicine, 4(10), 1547-1557 (2015-07-16)
Cancer cells exhibit a variety of features indicative of atypical nuclei. However, the molecular mechanisms underlying these phenomena remain to be elucidated. The linker of nucleoskeleton and cytoskeleton (LINC) complex, a nuclear envelope protein complex consisting mainly of the SUN
The leukocyte nuclear envelope proteome varies with cell activation and contains novel transmembrane proteins that affect genome architecture.
Korfali, N; Wilkie, GS; Swanson, SK; Srsen, V; Batrakou, DG; Fairley, EA; Malik et al.
Molecular and Cellular Proteomics null
Nadia Korfali et al.
Methods in molecular biology (Clifton, N.J.), 1411, 3-44 (2016-05-06)
Nuclei can be relatively easily extracted from homogenized liver due to the softness of the tissue and crudely separated from other cellular organelles by low-speed centrifugation due to the comparatively large size of nuclei. However, further purification is complicated by
Ayaka Matsumoto et al.
Methods in molecular biology (Clifton, N.J.), 1840, 307-319 (2018-08-25)
The linker of nucleoskeleton and cytoskeleton (LINC) complex, containing the proteins SUN and nesprin, is the fundamental structural unit of the nuclear envelope. The neoplastic-based regulation of the LINC complex in cancer tissues has become increasingly recognized in recent years
Nikolaj Zuleger et al.
Genome biology, 14(2), R14-R14 (2013-02-19)
Different cell types have distinctive patterns of chromosome positioning in the nucleus. Although ectopic affinity-tethering of specific loci can be used to relocate chromosomes to the nuclear periphery, endogenous nuclear envelope proteins that control such a mechanism in mammalian cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service