Skip to Content
Merck
All Photos(1)

Key Documents

MANH0C9B92E8

Manchester Organics

tert-Butyl 2,8-Diazaspiro[3.4]octane-8-carboxylate hemioxalate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
512.6
MDL number:

Iupac Name

tert-butyl 2,5-diazaspiro[3.4]octane-5-carboxylate, oxalate salt

Inchi Code

1S/2C11H20N2O2.C2H2O4/c2*1-10(2,3)15-9(14)13-6-4-5-11(13)7-12-8-11;3-1(4)2(5)6/h2*12H,4-8H2,1-3H3;(H,3,4)(H,5,6)/p-2

InChI key

HAGQSTKTUAYTMX-UHFFFAOYSA-L

Purity

95%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/H31909

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service