Skip to Content
Merck
All Photos(1)

Key Documents

MAN326368985

Manchester Organics

1-Benzyl-3-methyl-4-piperidone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
203.28
MDL number:

Iupac Name

1-benzyl-3-methyl-4-piperidinone

Inchi Code

1S/C13H17NO/c1-11-9-14(8-7-13(11)15)10-12-5-3-2-4-6-12/h2-6,11H,7-10H2,1H3

InChI key

OVQAJYCAXPHYNV-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/U23437

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service