ENAH99BD79DE
Enamine
1-chloro-1-fluoro-5-azaspiro[2.3]hexane hydrochloride
Select a Size
Vendor SKU: EN300-45657585-50MG
CZK 8,860.00
Select a Size
About This Item
Vendor SKU: EN300-45657585-50MG
CZK 8,860.00
Iupac Name
1-chloro-1-fluoro-5-azaspiro[2.3]hexane hydrochloride
Inchi Code
1S/C5H7ClFN.ClH/c6-5(7)1-4(5)2-8-3-4;/h8H,1-3H2;1H
InChI key
FHVKJVMEVNNMBG-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-45657585
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service