Skip to Content
Merck
All Photos(1)

Key Documents

ENAH93E74A1B

Enamine

methyl 5-bromo-2-cyclopropylpyridine-4-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 10,907.50

Vendor SKU: EN300-6490339-50MG

CZK 10,907.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 10,907.50

About This Item

CAS Number:
Molecular Weight:
256.1
MDL number:

Vendor SKU: EN300-6490339-50MG

CZK 10,907.50


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl 5-bromo-2-cyclopropylisonicotinate

Inchi Code

1S/C10H10BrNO2/c1-14-10(13)7-4-9(6-2-3-6)12-5-8(7)11/h4-6H,2-3H2,1H3

InChI key

JJZSVHBWGYNQRV-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6490339

MSDS

https://www.enaminestore.com/catalog/EN300-6490339


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service