Skip to Content
Merck
All Photos(1)

Key Documents

ENAH5802DABA

Enamine

{5-azaspiro[3.4]octan-7-yl}methanol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 7,800.00
100 MG
CZK 11,667.50
250 MG
CZK 16,672.50
500 MG
CZK 26,195.00
1 G
CZK 33,605.00
2.5 G
CZK 65,845.00
5 G
CZK 97,402.50
10 G
CZK 144,462.50

Vendor SKU: EN300-7425050-50MG

CZK 7,800.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 7,800.00
100 MG
CZK 11,667.50
250 MG
CZK 16,672.50
500 MG
CZK 26,195.00
1 G
CZK 33,605.00
2.5 G
CZK 65,845.00
5 G
CZK 97,402.50
10 G
CZK 144,462.50

About This Item

CAS Number:
Molecular Weight:
141.21
MDL number:

Vendor SKU: EN300-7425050-50MG

CZK 7,800.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

(5-azaspiro[3.4]octan-7-yl)methanol

Inchi Code

1S/C8H15NO/c10-6-7-4-8(9-5-7)2-1-3-8/h7,9-10H,1-6H2

InChI key

PDZDFXXMGUFNIQ-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-7425050

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-7425050


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service