Skip to Content
Merck
All Photos(1)

Key Documents

ENAH38A2D82E

Enamine

tert-butyl N-(4-amino-1,1-dioxo-1lambda6-thiolan-3-yl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 9,055.00
100 MG
CZK 12,955.00
250 MG
CZK 17,992.50
500 MG
CZK 27,677.50
1 G
CZK 35,152.50
2.5 G
CZK 67,847.50
5 G
CZK 99,827.50
10 G
CZK 147,472.50

Vendor SKU: EN300-6762704-50MG

CZK 9,055.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 9,055.00
100 MG
CZK 12,955.00
250 MG
CZK 17,992.50
500 MG
CZK 27,677.50
1 G
CZK 35,152.50
2.5 G
CZK 67,847.50
5 G
CZK 99,827.50
10 G
CZK 147,472.50

About This Item

CAS Number:
Molecular Weight:
250.32
MDL number:

Vendor SKU: EN300-6762704-50MG

CZK 9,055.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl (4-amino-1,1-dioxidotetrahydrothiophen-3-yl)carbamate

Inchi Code

1S/C9H18N2O4S/c1-9(2,3)15-8(12)11-7-5-16(13,14)4-6(7)10/h6-7H,4-5,10H2,1-3H3,(H,11,12)

InChI key

NNDORIZWDLLHLJ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-6762704

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-6762704


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service