Skip to Content
Merck
All Photos(1)

Key Documents

ENAH2C5577C9

Enamine

methyl cyclohex-2-ene-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 11,850.00
250 MG
CZK 16,432.50
500 MG
CZK 25,240.00
1 G
CZK 32,032.50
2.5 G
CZK 61,770.00
5 G
CZK 90,857.50
10 G
CZK 134,180.00

Vendor SKU: EN300-6237205-100MG

CZK 11,850.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 11,850.00
250 MG
CZK 16,432.50
500 MG
CZK 25,240.00
1 G
CZK 32,032.50
2.5 G
CZK 61,770.00
5 G
CZK 90,857.50
10 G
CZK 134,180.00

About This Item

CAS Number:
Molecular Weight:
140.18
MDL number:

Vendor SKU: EN300-6237205-100MG

CZK 11,850.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl cyclohex-2-ene-1-carboxylate

Inchi Code

1S/C8H12O2/c1-10-8(9)7-5-3-2-4-6-7/h3,5,7H,2,4,6H2,1H3

InChI key

LKVHTVRTQUBZDN-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-6237205

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service