Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11DD17B7

Enamine

rac-(1R,2S)-2-(trifluoromethyl)cyclopentan-1-amine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 17,310.00
100 MG
CZK 22,282.50
250 MG
CZK 31,382.50
500 MG
CZK 48,737.50
1 G
CZK 62,160.00
2.5 G
CZK 120,790.00

Vendor SKU: EN300-1716163-50MG

CZK 17,310.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 17,310.00
100 MG
CZK 22,282.50
250 MG
CZK 31,382.50
500 MG
CZK 48,737.50
1 G
CZK 62,160.00
2.5 G
CZK 120,790.00

About This Item

CAS Number:
Molecular Weight:
189.61
MDL number:

Vendor SKU: EN300-1716163-50MG

CZK 17,310.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

(1R,2S)-2-(trifluoromethyl)cyclopentan-1-amine hydrochloride

Inchi Code

1S/C6H10F3N.ClH/c7-6(8,9)4-2-1-3-5(4)10;/h4-5H,1-3,10H2;1H/t4-,5+;/m0./s1

InChI key

NNHZKEXLRJGUNR-UYXJWNHNSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1716163

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1716163


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service