Skip to Content
Merck
All Photos(1)

Key Documents

ENAH049F28F6

Enamine

2-chloro-5,6,7,8-tetrahydro-1,7-naphthyridine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 6,565.00
250 MG
CZK 9,392.50
500 MG
CZK 14,787.50
1 G
CZK 18,947.50
2.5 G
CZK 45,857.50
5 G
CZK 90,675.00
10 G
CZK 134,517.50

Vendor SKU: EN300-1694122-100MG

CZK 6,565.00


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 MG
CZK 6,565.00
250 MG
CZK 9,392.50
500 MG
CZK 14,787.50
1 G
CZK 18,947.50
2.5 G
CZK 45,857.50
5 G
CZK 90,675.00
10 G
CZK 134,517.50

About This Item

CAS Number:
Molecular Weight:
205.09
MDL number:

Vendor SKU: EN300-1694122-100MG

CZK 6,565.00


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

2-chloro-5,6,7,8-tetrahydro-1,7-naphthyridine hydrochloride

Inchi Code

1S/C8H9ClN2.ClH/c9-8-2-1-6-3-4-10-5-7(6)11-8;/h1-2,10H,3-5H2;1H

InChI key

HLJCOESZBRFFTH-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1694122

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1694122


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service