Skip to Content
Merck
All Photos(1)

Key Documents

ENAH0424E2AA

Enamine

methyl 1-(sulfamoylmethyl)cyclopropane-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 17,257.50
100 MG
CZK 22,555.00
250 MG
CZK 32,175.00
500 MG
CZK 50,700.00
1 G
CZK 65,032.50

Vendor SKU: EN300-1617668-50MG

CZK 17,257.50


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 MG
CZK 17,257.50
100 MG
CZK 22,555.00
250 MG
CZK 32,175.00
500 MG
CZK 50,700.00
1 G
CZK 65,032.50

About This Item

CAS Number:
Molecular Weight:
193.22
MDL number:

Vendor SKU: EN300-1617668-50MG

CZK 17,257.50


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

methyl 1-(sulfamoylmethyl)cyclopropane-1-carboxylate

Inchi Code

1S/C6H11NO4S/c1-11-5(8)6(2-3-6)4-12(7,9)10/h2-4H2,1H3,(H2,7,9,10)

InChI key

DAPTVWLENMABTN-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1617668

MSDS

https://www.enaminestore.com/catalog/EN300-1617668


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service