Skip to Content
Merck
All Photos(1)

Key Documents

ENAH03BC21D6

Enamine

4-{[2-(prop-2-yn-1-ylsulfanyl)ethyl]amino}pyrimidine-5-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 11,362.50

Vendor SKU: EN300-1449931-50MG

CZK 11,362.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 11,362.50

About This Item

CAS Number:
Molecular Weight:
237.28
MDL number:

Vendor SKU: EN300-1449931-50MG

CZK 11,362.50


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-((2-(prop-2-yn-1-ylthio)ethyl)amino)pyrimidine-5-carboxylic acid

Inchi Code

1S/C10H11N3O2S/c1-2-4-16-5-3-12-9-8(10(14)15)6-11-7-13-9/h1,6-7H,3-5H2,(H,14,15)(H,11,12,13)

InChI key

AUKMBNSXJCXPHJ-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1449931

MSDS

https://www.enaminestore.com/catalog/EN300-1449931


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service