Skip to Content
Merck
All Photos(1)

Key Documents

ENAH03855EBA

Enamine

3-methyl-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine-6-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 9,087.50
250 MG
CZK 12,500.00
500 MG
CZK 19,032.50
1 G
CZK 24,102.50
2.5 G
CZK 46,137.50
5 G
CZK 67,717.50

Vendor SKU: EN300-259830-100MG

CZK 9,087.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 9,087.50
250 MG
CZK 12,500.00
500 MG
CZK 19,032.50
1 G
CZK 24,102.50
2.5 G
CZK 46,137.50
5 G
CZK 67,717.50

About This Item

CAS Number:
Molecular Weight:
181.2
MDL number:

Vendor SKU: EN300-259830-100MG

CZK 9,087.50


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-methyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine-6-carboxamide

Inchi Code

1S/C7H11N5O/c1-4-10-11-6-2-9-5(7(8)13)3-12(4)6/h5,9H,2-3H2,1H3,(H2,8,13)

InChI key

HXWSCHMRBNKAEU-UHFFFAOYSA-N

storage temp.

RT

Purity

91

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-259830

MSDS

https://www.enaminestore.com/catalog/EN300-259830


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service