Skip to Content
Merck
All Photos(1)

Key Documents

ENA964506784

Enamine

4,5-difluoro-2-methoxybenzene-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 8,385.00
250 MG
CZK 12,057.50
500 MG
CZK 18,947.50
1 G
CZK 24,310.00
2.5 G
CZK 47,612.50
5 G
CZK 70,460.00

Vendor SKU: EN300-738605-100MG

CZK 8,385.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 8,385.00
250 MG
CZK 12,057.50
500 MG
CZK 18,947.50
1 G
CZK 24,310.00
2.5 G
CZK 47,612.50
5 G
CZK 70,460.00

About This Item

CAS Number:
Molecular Weight:
242.63
MDL number:

Vendor SKU: EN300-738605-100MG

CZK 8,385.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

4,5-difluoro-2-methoxybenzenesulfonyl chloride

Inchi Code

1S/C7H5ClF2O3S/c1-13-6-2-4(9)5(10)3-7(6)14(8,11)12/h2-3H,1H3

InChI key

MARRIMPKLITFCM-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-738605

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-738605


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service