Skip to Content
Merck
All Photos(1)

Key Documents

ENA638598870

Enamine

1-tert-butyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 17,700.00
100 MG
CZK 22,835.00
250 MG
CZK 32,032.50
500 MG
CZK 49,842.50
1 G
CZK 63,622.50
2.5 G
CZK 123,617.50
5 G
CZK 182,345.00
10 G
CZK 269,770.00

Vendor SKU: EN300-344916-50MG

CZK 17,700.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 17,700.00
100 MG
CZK 22,835.00
250 MG
CZK 32,032.50
500 MG
CZK 49,842.50
1 G
CZK 63,622.50
2.5 G
CZK 123,617.50
5 G
CZK 182,345.00
10 G
CZK 269,770.00

About This Item

CAS Number:
Molecular Weight:
195.22
MDL number:

Vendor SKU: EN300-344916-50MG

CZK 17,700.00


Ships in 14 days - from Aldrich Partner.

mp

129-132

Iupac Name

1-tert-butyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid

Inchi Code

1S/C10H13NO3/c1-10(2,3)11-6-4-5-7(8(11)12)9(13)14/h4-6H,1-3H3,(H,13,14)

InChI key

ODRSRNLRTYDDRX-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-344916

Melting Point

129-132

MSDS

https://www.enaminestore.com/catalog/EN300-344916


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service